Tisk…

N-[1-(4-Fluorophenyl)ethyl]cyclopropanamine hydrochloride

Dodavatel: Apollo Scientific
Varování

PC512015-1G PC512015-250MG
APOSPC512015-1GEA 2980 CZK
APOSPC512015-1G APOSPC512015-250MG
N-[1-(4-Fluorophenyl)ethyl]cyclopropanamine hydrochloride
N-[1-(4-Fluorophenyl)ethyl]cyclopropanamine hydrochloride
Vzorec: C₁₁H₁₄FN·ClH
MW: 215.695 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD31695784
CAS číslo: 2366994-30-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR