Tisk…

N-[(1S)-1-(4-Fluorophenyl)ethyl]cyclopropanamine hydrochloride

Dodavatel: Apollo Scientific
Varování

PC512014-1G PC512014-250MG
APOSPC512014-1GEA 5920 CZK
APOSPC512014-1G APOSPC512014-250MG
N-[(1S)-1-(4-Fluorophenyl)ethyl]cyclopropanamine hydrochloride
N-[(1S)-1-(4-Fluorophenyl)ethyl]cyclopropanamine hydrochloride
Vzorec: C₁₁H₁₄FN·ClH
MW: 215,695 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31695606
CAS číslo: 2407312-04-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR