Tisk…

4-Bromo-6-(pentafluorothio)-1H-benzimidazole 95%

Dodavatel: Apollo Scientific
Varování

Synonyms: (4-Bromo-1H-benzimidazol-6-yl)sulphur pentafluoride, 4-Bromo-6-(pentafluorosulphanyl)-1H-benzimidazole

PC51190-1G PC51190-250MG
APOSPC51190-1GEA 12200 CZK
APOSPC51190-1G APOSPC51190-250MG
4-Bromo-6-(pentafluorothio)-1H-benzimidazole 95%
4-Bromo-6-(pentafluorothio)-1H-benzimidazole
Vzorec: C₇H₄BrF₅N₂S
MW: 323,081 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24369706
CAS číslo: 1432075-84-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR