5-Bromo-4-fluoro-2-hydroxybenzoic acid 96%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
4-Bromo-2-carboxy-5-fluorophenol, 5-Bromo-4-fluorosalicylic acid
PC51184-1G
PC51184-250MG
PC51184-5G
APOSPC51184-1GEA
5960
CZK
APOSPC51184-1G
APOSPC51184-250MG
APOSPC51184-5G
5-Bromo-4-fluoro-2-hydroxybenzoic acid 96%
5-Bromo-4-fluoro-2-hydroxybenzoic acid
Vzorec:
C₇H₄BrFO₃ MW: 235,007 g/mol Bod tání: 203…208 °C Storage Temperature: Ambient |
MDL Number:
MFCD08235149 CAS číslo: 1644-71-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...