2,4-Difluoro-3-(trifluoromethyl)benzaldehyde 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
α,α,α,2,4-Pentafluoro-m-tolualdehyde, 2,6-Difluoro-3-formylbenzotrifluoride
PC51182-1G
PC51182-250MG
PC51182-5G
APOSPC51182-1GEA
2860
CZK
APOSPC51182-1G
APOSPC51182-250MG
APOSPC51182-5G
2,4-Difluoro-3-(trifluoromethyl)benzaldehyde 98%
2,4-Difluoro-3-(trifluoromethyl)benzaldehyde
Vzorec:
C₈H₃F₅O MW: 210.101 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD12404958 CAS číslo: 134099-30-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...