2'-Fluoro-3'-(trifluoromethoxy)acetophenone 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
1-[2-Fluoro-3-(trifluoromethoxy)phenyl]ethan-1-one
PC51175-1G
PC51175-250MG
PC51175-5G
APOSPC51175-1GEA
4470
CZK
APOSPC51175-1G
APOSPC51175-250MG
APOSPC51175-5G
2'-Fluoro-3'-(trifluoromethoxy)acetophenone 97%
2'-Fluoro-3'-(trifluoromethoxy)acetophenone
Vzorec:
C₉H₆F₄O₂ Storage Temperature: Ambient |
MDL Number:
MFCD23703636 CAS číslo: 1429321-84-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...