Tisk…

2-Amino-4-[(trifluoromethyl)thio]phenol 95%

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-Amino-4-[(trifluoromethyl)sulphanyl]phenol, 2-Hydroxy-5-[(trifluoromethyl)thio]aniline

PC51173-250MG PC51173-1G
APOSPC51173-250MGEA 3230 CZK
APOSPC51173-250MG APOSPC51173-1G
2-Amino-4-[(trifluoromethyl)thio]phenol 95%
2-Amino-4-[(trifluoromethyl)thio]phenol
Vzorec: C₇H₆F₃NOS
MW: 209,189 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD23703635
CAS číslo: 228401-48-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR