Tisk…

4-Bromo-2,3-difluorobenzyl bromide 97%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: α,4-Dibromo-2,3-difluorotoluene, 1-Bromo-4-(bromomethyl)-2,3-difluorobenzene

PC51172-1G PC51172-250MG PC51172-5G
APOSPC51172-1GEA 8900 CZK
APOSPC51172-1G APOSPC51172-250MG APOSPC51172-5G
4-Bromo-2,3-difluorobenzyl bromide 97%
4-Bromo-2,3-difluorobenzyl bromide
Vzorec: C₇H₄Br₂F₂
MW: 285.911 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23703634
CAS číslo: 162744-56-1
UN: 3265
ADR: 8 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR