Tisk…

2-Bromo-6-nitro-4-[(trifluoromethyl)thio]phenol

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-Bromo-6-nitro-4-[(trifluoromethyl)sulphanyl]phenol, 3-Bromo-4-hydroxy-5-nitrophenyl trifluoromethyl sulphide

PC51171-1G PC51171-250MG PC51171-5G
APOSPC51171-1GEA 19510 CZK
APOSPC51171-1G APOSPC51171-250MG APOSPC51171-5G
2-Bromo-6-nitro-4-[(trifluoromethyl)thio]phenol
2-Bromo-6-nitro-4-[(trifluoromethyl)thio]phenol
Vzorec: C₇H₃BrF₃NO₃S
MW: 318,068 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23703633
CAS číslo: 1429321-81-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR