Tisk…

Perfluoro-5-(fluorosulphonyl)-2-methyl-3-oxapentanoyl fluoride

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 5-(Fluorosulphonyl)-2,4,4,5,5-pentafluoro-2-(trifluoromethyl)-3-oxapentanoyl fluoride, 2,3,3,3-Tetrafluoro-2-[1,1,2,2-tetrafluoro-3-(fluorosulphonyl)ethoxy]propanoyl fluoride

PC51168-1G PC51168-25G PC51168-5G
APOSPC51168-1GEA 3740 CZK
APOSPC51168-1G APOSPC51168-25G APOSPC51168-5G
Perfluoro-5-(fluorosulphonyl)-2-methyl-3-oxapentanoyl fluoride
Perfluoro-5-(fluorosulphonyl)-2-methyl-3-oxapentanoyl fluoride
Vzorec: C₅F₁₀O₄S
Storage Temperature: Ambient
MDL Number: MFCD16620593
CAS číslo: 4089-57-0
UN: 3265
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR