Tisk…

2-Nitro-4-[(trifluoromethyl)thio]phenol 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: 4-Hydroxy-3-nitrophenyl trifluoromethyl sulphide, 2-Nitro-4-[(trifluoromethyl)sulphanyl]phenol, 2-Nitro-4-((trifluoromethyl)thio)phenol

PC51167-1G PC51167-250MG PC51167-5G
APOSPC51167-1GEA 1240 CZK
APOSPC51167-1G APOSPC51167-250MG APOSPC51167-5G
2-Nitro-4-[(trifluoromethyl)thio]phenol 97%
2-Nitro-4-[(trifluoromethyl)thio]phenol
Vzorec: C₇H₄F₃NO₃S
MW: 239,172 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08457463
CAS číslo: 933673-33-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR