Tisk…

4-Bromo-3-fluoro-2-hydroxybenzoic acid 97%

Dodavatel: Apollo Scientific
Varování

PC51165-1G PC51165-250MG
APOSPC51165-1GEA 0 CZK
APOSPC51165-1G APOSPC51165-250MG
4-Bromo-3-fluoro-2-hydroxybenzoic acid 97%
4-Bromo-3-fluoro-2-hydroxybenzoic acid
Vzorec: C₇H₄BrFO₃
Storage Temperature: Refrigerator
MDL Number: MFCD23703631
CAS číslo: 1429321-82-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR