Tisk…

2,3-Difluoro-4-hydroxypyridine 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: 2,3-Difluoropyridin-4-ol

PC51124-1G PC51124-500MG
APOSPC51124-1GEA 12870 CZK
APOSPC51124-1G APOSPC51124-500MG
2,3-Difluoro-4-hydroxypyridine 98%
2,3-Difluoro-4-hydroxypyridine
Vzorec: C₅H₃F₂NO
MW: 131,08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16610798
CAS číslo: 1227579-00-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR