Tisk…

2-Bromo-3-iodobenzotrifluoride 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-Bromo-1-iodo-3-(trifluoromethyl)benzene, 2-Bromo-3-iodo-α,α,α-trifluorotoluene

PC51105-100MG PC51105-1G PC51105-250MG
APOSPC51105-100MGEA 4070 CZK
APOSPC51105-100MG APOSPC51105-1G APOSPC51105-250MG
2-Bromo-3-iodobenzotrifluoride 97%
2-Bromo-3-iodobenzotrifluoride
Vzorec: C₇H₃BrF₃I
MW: 350,902 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD17012838
CAS číslo: 1049731-01-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR