5-(Trifluoromethyl)pyridine-2,3-diamine 98%
Dodavatel: Apollo Scientific
Synonyms:
2,3-Diamino-5-(trifluoromethyl)pyridine
PC51080-100MG
PC51080-1G
PC51080-250MG
APOSPC51080-100MGEA
4100
CZK
APOSPC51080-100MG
APOSPC51080-1G
APOSPC51080-250MG
5-(Trifluoromethyl)pyridine-2,3-diamine 98%
5-(Trifluoromethyl)pyridine-2,3-diamine
Vzorec:
C₆H₆F₃N₃ MW: 177,127 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD10696252 CAS číslo: 107867-51-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...