1,2,3,4,5-Pentafluoro-6-(prop-1-en-1-yl)benzene
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
1-(Perfluorophenyl)prop-1-ene, 1-(Pentafluorophenyl)prop-1-ene
PC51069-1G
PC51069-5G
APOSPC51069-1GEA
4070
CZK
APOSPC51069-1G
APOSPC51069-5G
1,2,3,4,5-Pentafluoro-6-(prop-1-en-1-yl)benzene
1,2,3,4,5-Pentafluoro-6-(prop-1-en-1-yl)benzene
Vzorec:
C₉H₅F₅ MW: 208,128 g/mol Bod varu: 154…162 °C Storage Temperature: Ambient |
MDL Number:
MFCD20481819 CAS číslo: 15948-70-6 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...