Tisk…

Perfluoro(chlorocyclopentane) 90%

Dodavatel: Apollo Scientific
Varování

Synonyms: 1-Chloro-1,2,2,3,3,4,4,5,5-nonafluorocyclopentane

PC51062-1G PC51062-250MG
APOSPC51062-1GEA 12960 CZK
APOSPC51062-1G APOSPC51062-250MG
Perfluoro(chlorocyclopentane) 90%
Perfluoro(chlorocyclopentane)
Vzorec: C₅ClF₉
Bod varu: 52 °C
Hustota: 1,61 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD19442028
CAS číslo: 376-76-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR