Tisk…

Sodium 5H-octafluoropentanoate

Dodavatel: Apollo Scientific

PC51057-1G PC51057-5G
APOSPC51057-1GEA 1660 CZK
APOSPC51057-1G APOSPC51057-5G
Sodium 5H-octafluoropentanoate
Sodium 5H-octafluoropentanoate
Vzorec: C₅HF₈O₂·Na
Bod tání: 239…241 °C
Storage Temperature: Ambient
MDL Number: MFCD00188425
CAS číslo: 22715-46-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR