Tisk…

2-(2,2-Difluoroethoxy)pyrimidine-5-carbaldehyde 95%

Dodavatel: Apollo Scientific

PC510359-1G PC510359-250MG
APOSPC510359-1GEA 31740 CZK
APOSPC510359-1G APOSPC510359-250MG
2-(2,2-Difluoroethoxy)pyrimidine-5-carbaldehyde 95%
2-(2,2-Difluoroethoxy)pyrimidine-5-carbaldehyde
Vzorec: C₇H₆F₂N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD26843607
CAS číslo: 1565712-50-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR