Tisk…

6-Amino-5-fluoropyridine-3-boronic acid pinacol ester 97%

Dodavatel: Apollo Scientific

Synonyms: 3-Fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-amine

PC510356-1G PC510356-250MG
APOSPC510356-1GEA 13860 CZK
APOSPC510356-1G APOSPC510356-250MG
6-Amino-5-fluoropyridine-3-boronic acid pinacol ester 97%
6-Amino-5-fluoropyridine-3-boronic acid pinacol ester
Vzorec: C₁₁H₁₆BFN₂O₂
MW: 238.066 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12923417
CAS číslo: 944401-75-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR