2-Amino-5-(trifluoromethoxy)benzimidazole 95%
Dodavatel: Apollo Scientific
PC510355-1G
PC510355-250MG
APOSPC510355-1GEA
22220
CZK
APOSPC510355-1G
APOSPC510355-250MG
2-Amino-5-(trifluoromethoxy)benzimidazole 95%
2-Amino-5-(trifluoromethoxy)benzimidazole
Vzorec:
C₈H₆F₃N₃O MW: 217,148 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07778330 CAS číslo: 132877-27-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...