4-(Boc-Amino)-2-fluoro-5-nitrobenzonitrile 97%
Dodavatel: Apollo Scientific
PC510352-1G
PC510352-250MG
PC510352-5G
APOSPC510352-1GEA
15150
CZK
APOSPC510352-1G
APOSPC510352-250MG
APOSPC510352-5G
4-(Boc-Amino)-2-fluoro-5-nitrobenzonitrile 97%
4-(Boc-Amino)-2-fluoro-5-nitrobenzonitrile
Vzorec:
C₁₂H₁₂FN₃O₄ MW: 281,24 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD31695792 CAS číslo: 473537-38-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...