2-Amino-6-chloro-3-fluorobenzoic acid
Dodavatel: Apollo Scientific
PC510347-1G
PC510347-250MG
APOSPC510347-1GEA
5060
CZK
APOSPC510347-1G
APOSPC510347-250MG
2-Amino-6-chloro-3-fluorobenzoic acid
Vzorec:
C₇H₅ClFNO₂ Storage Temperature: Ambient |
MDL Number:
MFCD11183542 CAS číslo: 1039815-76-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...