Tisk…

Ethyl 2,6-Dichloro-5-fluoropyrimidine-4-carboxylate

Dodavatel: Apollo Scientific

PC510346-1G PC510346-250MG
APOSPC510346-1GEA 31910 CZK
APOSPC510346-1G APOSPC510346-250MG
Ethyl 2,6-Dichloro-5-fluoropyrimidine-4-carboxylate
Ethyl 2,6-Dichloro-5-fluoropyrimidine-4-carboxylate
Vzorec: C₇H₅Cl₂FN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD29065911
CAS číslo: 1266238-63-4

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR