4-(Carboxydifluoromethyl)benzoic acid
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC510332-1G
PC510332-250MG
APOSPC510332-1GEA
5630
CZK
APOSPC510332-1G
APOSPC510332-250MG
4-(Carboxydifluoromethyl)benzoic acid
4-(Carboxydifluoromethyl)benzoic acid
Vzorec:
C₉H₆F₂O₄ Storage Temperature: Ambient |
MDL Number:
MFCD21099440 CAS číslo: 1240313-90-9 UN: 1759 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...