(R)-4-Amino-3-fluoro-2-methyl-2-butanol
Dodavatel: Apollo Scientific
|
Varování
|
PC510301-1G
PC510301-250MG
PC510301-5G
APOSPC510301-1GEA
12770
CZK
APOSPC510301-1G
APOSPC510301-250MG
APOSPC510301-5G
(R)-4-Amino-3-fluoro-2-methyl-2-butanol
(R)-4-Amino-3-fluoro-2-methyl-2-butanol
Vzorec:
C₅H₁₂FNO MW: 121,153 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD29055294 CAS číslo: 1544241-64-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...