Tisk…

5-(2-Furyl)-3-(trifluoromethyl)pyrazole

Dodavatel: Apollo Scientific

PC510294-1G PC510294-5G
APOSPC510294-1GEA 6980 CZK
APOSPC510294-1G APOSPC510294-5G
5-(2-Furyl)-3-(trifluoromethyl)pyrazole
5-(2-Furyl)-3-(trifluoromethyl)pyrazole
Vzorec: C₈H₅F₃N₂O
Storage Temperature: Ambient
MDL Number: MFCD07367235
CAS číslo: 197507-85-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR