Methyl 2-Fluoro-6-methoxy-3-(methylsulphonyl)benzoate
Dodavatel: Apollo Scientific
PC510292-1G
PC510292-5G
APOSPC510292-1GEA
11310
CZK
APOSPC510292-1G
APOSPC510292-5G
Methyl 2-Fluoro-6-methoxy-3-(methylsulphonyl)benzoate
Methyl 2-Fluoro-6-methoxy-3-(methylsulphonyl)benzoate
Vzorec:
C₁₀H₁₁FO₅S MW: 262,255 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD29049236 CAS číslo: 2006278-07-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...