Tisk…

Ethyl 2-(3-Chloro-4-fluorophenyl)acetate

Dodavatel: Apollo Scientific

PC510288-1G PC510288-5G
APOSPC510288-1GEA 8700 CZK
APOSPC510288-1G APOSPC510288-5G
Ethyl 2-(3-Chloro-4-fluorophenyl)acetate
Ethyl 2-(3-Chloro-4-fluorophenyl)acetate
Vzorec: C₁₀H₁₀ClFO₂
MW: 216,637 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11045165
CAS číslo: 340825-20-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR