Tisk…

2-Fluoro-1-iodo-2-methylpropane

Dodavatel: Apollo Scientific

PC510283-1G PC510283-250MG
APOSPC510283-1GEA 28700 CZK
APOSPC510283-1G APOSPC510283-250MG
2-Fluoro-1-iodo-2-methylpropane
2-Fluoro-1-iodo-2-methylpropane
Vzorec: C₄H₈FI
MW: 202,009 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24458933
CAS číslo: 19869-79-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR