Tisk…

Methyl (E)-3-[2-Amino-4-(trifluoromethyl)phenyl]acrylate

Dodavatel: Apollo Scientific

PC510279-1G PC510279-250MG PC510279-5G
APOSPC510279-1GEA 14060 CZK
APOSPC510279-1G APOSPC510279-250MG APOSPC510279-5G
Methyl (E)-3-[2-Amino-4-(trifluoromethyl)phenyl]acrylate
Methyl (E)-3-[2-Amino-4-(trifluoromethyl)phenyl]acrylate
Vzorec: C₁₁H₁₀F₃NO₂
MW: 245,198 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28678072
CAS číslo: 231296-89-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR