Tisk…

3-Ethoxy-2-fluoropyridine

Dodavatel: Apollo Scientific

PC510249-1G PC510249-250MG PC510249-5G
APOSPC510249-1GEA 22620 CZK
APOSPC510249-1G APOSPC510249-250MG APOSPC510249-5G
3-Ethoxy-2-fluoropyridine
3-Ethoxy-2-fluoropyridine
Vzorec: C₇H₈FNO
MW: 141,143 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18384511
CAS číslo: 847225-55-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR