Tisk…

2-[4-(Trifluoromethoxy)phenyl]thiazole-5-carboxylic acid

Dodavatel: Apollo Scientific

PC510243-1G PC510243-250MG PC510243-5G
APOSPC510243-1GEA 14220 CZK
APOSPC510243-1G APOSPC510243-250MG APOSPC510243-5G
2-[4-(Trifluoromethoxy)phenyl]thiazole-5-carboxylic acid
2-[4-(Trifluoromethoxy)phenyl]thiazole-5-carboxylic acid
Vzorec: C₁₁H₆F₃NO₃S
MW: 289,23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07375334
CAS číslo: 886370-20-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR