Tisk…

3-(3-Fluoro-4-methoxyphenyl)propionic acid

Dodavatel: Apollo Scientific

PC510235-1G PC510235-5G
APOSPC510235-1GEA 3410 CZK
APOSPC510235-1G APOSPC510235-5G
3-(3-Fluoro-4-methoxyphenyl)propionic acid
3-(3-Fluoro-4-methoxyphenyl)propionic acid
Vzorec: C₁₀H₁₁FO₃
MW: 198,191 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06823839
CAS číslo: 69888-90-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR