2-[3,5-Bis(trifluoromethyl)phenyl]oxirane
Dodavatel: Apollo Scientific
PC510217-1G
PC510217-250MG
APOSPC510217-1GEA
17130
CZK
APOSPC510217-1G
APOSPC510217-250MG
2-[3,5-Bis(trifluoromethyl)phenyl]oxirane
2-[3,5-Bis(trifluoromethyl)phenyl]oxirane
Vzorec:
C₁₀H₆F₆O MW: 256,144 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16503160 CAS číslo: 109086-18-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...