Tisk…

3,3,3-Trifluoro-2-phenylpropionic acid

Dodavatel: Apollo Scientific

PC510212-1G PC510212-250MG
APOSPC510212-1GEA 55220 CZK
APOSPC510212-1G APOSPC510212-250MG
3,3,3-Trifluoro-2-phenylpropionic acid
3,3,3-Trifluoro-2-phenylpropionic acid
Vzorec: C₉H₇F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD16616839
CAS číslo: 56539-85-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR