Tisk…

3-Fluoro-4-methoxyphenethyl Alcohol

Dodavatel: Apollo Scientific

Synonyms: 2-(3-Fluoro-4-methoxyphenyl)ethanol

PC510207-5G PC510207-1G
APOSPC510207-5GEA 7440 CZK
APOSPC510207-5G APOSPC510207-1G
3-Fluoro-4-methoxyphenethyl Alcohol
3-Fluoro-4-methoxyphenethyl Alcohol
Vzorec: C₉H₁₁FO₂
MW: 170,181 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06201038
CAS číslo: 404-91-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR