2-Methoxy-5-nitro-3-(trifluoromethyl)pyridine
Dodavatel: Apollo Scientific
PC510197-1G
PC510197-250MG
APOSPC510197-1GEA
17130
CZK
APOSPC510197-1G
APOSPC510197-250MG
2-Methoxy-5-nitro-3-(trifluoromethyl)pyridine
2-Methoxy-5-nitro-3-(trifluoromethyl)pyridine
Vzorec:
C₇H₅F₃N₂O₃ MW: 222,121 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD17168448 CAS číslo: 1803857-10-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...