4-Bromo-2-fluorophenylboronic acid pinacol ester
Dodavatel: Apollo Scientific
PC510146-5G
PC510146-1G
APOSPC510146-5GEA
3810
CZK
APOSPC510146-5G
APOSPC510146-1G
4-Bromo-2-fluorophenylboronic acid pinacol ester
4-Bromo-2-fluorophenylboronic acid pinacol ester
Vzorec:
C₁₂H₁₅BBrFO₂ Storage Temperature: Ambient |
MDL Number:
MFCD22494116 CAS číslo: 1326316-85-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...