Tisk…

5-Bromo-3-(trifluoroacetyl)indole

Dodavatel: Apollo Scientific

Synonyms: 1-(5-Bromo-1H-indol-3-yl)-2,2,2-trifluoroethanone

PC510142-5G PC510142-1G
APOSPC510142-5GEA 11310 CZK
APOSPC510142-5G APOSPC510142-1G
5-Bromo-3-(trifluoroacetyl)indole
5-Bromo-3-(trifluoroacetyl)indole
Vzorec: C₁₀H₅BrF₃NO
MW: 292,052 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03721051
CAS číslo: 32387-18-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR