Ethyl 2-Fluorothiazole-4-carboxylate
Dodavatel: Apollo Scientific
PC510136-1G
PC510136-250MG
APOSPC510136-1GEA
8730
CZK
APOSPC510136-1G
APOSPC510136-250MG
Ethyl 2-Fluorothiazole-4-carboxylate
Ethyl 2-Fluorothiazole-4-carboxylate
Vzorec:
C₆H₆FNO₂S Storage Temperature: Ambient |
MDL Number:
MFCD18416559 CAS číslo: 153027-86-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...