Tisk…

Diethyl 2-Fluorobenzylphosphonate

Dodavatel: Apollo Scientific

PC510133-5G PC510133-1G
APOSPC510133-5GEA 3640 CZK
APOSPC510133-5G APOSPC510133-1G
Diethyl 2-Fluorobenzylphosphonate
Diethyl 2-Fluorobenzylphosphonate
Vzorec: C₁₁H₁₆FO₃P
Storage Temperature: Ambient
MDL Number: MFCD22574982
CAS číslo: 63909-54-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR