Tisk…

3,4-Difluoro-N-(6-nitro-2-benzothiazolyl)benzamide

Dodavatel: Apollo Scientific

PC510107-1G PC510107-250MG
APOSPC510107-1GEA 17130 CZK
APOSPC510107-1G APOSPC510107-250MG
3,4-Difluoro-N-(6-nitro-2-benzothiazolyl)benzamide
3,4-Difluoro-N-(6-nitro-2-benzothiazolyl)benzamide
Vzorec: C₁₄H₇F₂N₃O₃S
MW: 335,285 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00987073
CAS číslo: 313222-96-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR