Tisk…

Ethyl 2-(3-Fluorophenoxy)acetate

Dodavatel: Apollo Scientific

PC510104-1G PC510104-5G
APOSPC510104-1GEA 3540 CZK
APOSPC510104-1G APOSPC510104-5G
Ethyl 2-(3-Fluorophenoxy)acetate
Ethyl 2-(3-Fluorophenoxy)acetate
Vzorec: C₁₀H₁₁FO₃
MW: 198,191 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06659702
CAS číslo: 777-70-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR