5-[3,5-Bis(trifluoromethyl)phenyl]-3-pyridinecarbaldehyde
Dodavatel: Apollo Scientific
PC510097-5G
PC510097-1G
APOSPC510097-5GEA
40340
CZK
APOSPC510097-5G
APOSPC510097-1G
5-[3,5-Bis(trifluoromethyl)phenyl]-3-pyridinecarbaldehyde
5-[3,5-Bis(trifluoromethyl)phenyl]-3-pyridinecarbaldehyde
Vzorec:
C₁₄H₇F₆NO MW: 319,202 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06410252 CAS číslo: 887973-75-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...