Tisk…

6-[4-(Trifluoromethoxy)phenyl]-2-pyridinecarbaldehyde

Dodavatel: Apollo Scientific

PC510090-1G PC510090-5G PC510090-250MG
APOSPC510090-1GEA 14220 CZK
APOSPC510090-1G APOSPC510090-5G APOSPC510090-250MG
6-[4-(Trifluoromethoxy)phenyl]-2-pyridinecarbaldehyde
6-[4-(Trifluoromethoxy)phenyl]-2-pyridinecarbaldehyde
Vzorec: C₁₃H₈F₃NO₂
MW: 267,203 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06410031
CAS číslo: 887979-25-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR