Tisk…

6-[2-(Trifluoromethoxy)phenyl]-2-pyridinecarboxylic acid

Dodavatel: Apollo Scientific

PC510089-1G PC510089-5G
APOSPC510089-1GEA 12770 CZK
APOSPC510089-1G APOSPC510089-5G
6-[2-(Trifluoromethoxy)phenyl]-2-pyridinecarboxylic acid
6-[2-(Trifluoromethoxy)phenyl]-2-pyridinecarboxylic acid
Vzorec: C₁₃H₈F₃NO₃
MW: 283,203 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06410072
CAS číslo: 887983-48-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR