Tisk…

4-(Methylsulphonamido)-2-(trifluoromethyl)benzoic acid

Dodavatel: Apollo Scientific

PC510055-1G PC510055-5G
APOSPC510055-1GEA 8700 CZK
APOSPC510055-1G APOSPC510055-5G
4-(Methylsulphonamido)-2-(trifluoromethyl)benzoic acid
4-(Methylsulphonamido)-2-(trifluoromethyl)benzoic acid
Vzorec: C₉H₈F₃NO₄S
MW: 283.224 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19442791
CAS číslo: 1314406-51-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR