Tisk…

4-[(Cyclopropylcarbonyl)amino]-2-(trifluoromethyl)benzoic acid

Dodavatel: Apollo Scientific

PC510052-1G PC510052-5G
APOSPC510052-1GEA 8700 CZK
APOSPC510052-1G APOSPC510052-5G
4-[(Cyclopropylcarbonyl)amino]-2-(trifluoromethyl)benzoic acid
4-[(Cyclopropylcarbonyl)amino]-2-(trifluoromethyl)benzoic acid
Vzorec: C₁₂H₁₀F₃NO₃
MW: 273,208 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19442790
CAS číslo: 1314406-50-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR