2-[(Cyclopropylcarbonyl)amino]-5-fluorobenzoic acid
Dodavatel: Apollo Scientific
PC510051-1G
PC510051-5G
APOSPC510051-1GEA
7840
CZK
APOSPC510051-1G
APOSPC510051-5G
2-[(Cyclopropylcarbonyl)amino]-5-fluorobenzoic acid
2-[(Cyclopropylcarbonyl)amino]-5-fluorobenzoic acid
Vzorec:
C₁₁H₁₀FNO₃ MW: 223,2 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09935518 CAS číslo: 1016869-97-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...